4-Aminotetrahydropyran

Lieferant: FLUOROCHEM

Synonyme: Tetrahydro-2H-pyran-4-amine

023514
FLUO023514EA 0 EUR
FLUO023514
4-Aminotetrahydropyran
4-Aminotetrahydropyran
Formel: C₅H₁₁NO
Molecular Weight: 101,1469 g/mol
MDL: MFCD02179436
CAS-Nummer: 38041-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR