4-Methyl-2-thiophencarbonitril

Lieferant: FLUOROCHEM

023897-1G
FLUO023897-1GEA 495 EUR
FLUO023897-1G
4-Methyl-2-thiophencarbonitril
4-Methyl-2-thiophencarbonitril
Formel: C₆H₅NS
MDL: MFCD06657972
CAS-Nummer: 73305-93-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR