(S)-(+)-Epichlorhydrin
Lieferant: FLUOROCHEM
Synonyme:
(S)-(+)-3-Chlor-1,2-epoxypropan, (S)-(+)-Glycerolepichlorhydrin, (S)-(+)-1-Chlor-2,3-epoxypropan
023907-25G
023907-5G
FLUO023907-25GEA
121
EUR
FLUO023907-25G
FLUO023907-5G
(S)-(+)-Epichlorhydrin
(S)-(+)-Epichlorhydrin
Formel:
ClCH₂CH(O)CH₂ Molecular Weight: 92.52 g/mol Siedepunkt: 113…115 °C (1013 hPa) Schmelzpunkt: –48 °C Dichte: 1.18 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00077760 CAS-Nummer: 67843-74-7 UN: 2023 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...