N,O-Dimethylhydroxylamin Hydrochlorid

Lieferant: FLUOROCHEM

023913-1KG 023913-100G 023913-25G
FLUO023913-1KGEA 0 EUR
FLUO023913-1KG FLUO023913-100G FLUO023913-25G
N,O-Dimethylhydroxylamin Hydrochlorid
N,O-Dimethylhydroxylamin Hydrochlorid
Formel: C₂H₈ClNO
MDL: MFCD00012485
CAS-Nummer: 6638-79-5
EINECS: 229-642-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR