2-Amino-5-brom-6-fluorbenzonitril

Lieferant: FLUOROCHEM

Synonyme: 6-Amino-3-bromo-2-fluorobenzonitrile

023916-25G 023916-5G
FLUO023916-25GEA 705 EUR
FLUO023916-25G FLUO023916-5G
2-Amino-5-brom-6-fluorbenzonitril
2-Amino-5-brom-6-fluorbenzonitril
Formel: C₇H₄BrFN₂
Molecular Weight: 215.0225 g/mol
MDL: MFCD06657979
CAS-Nummer: 845866-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR