2-Chlor-4-phenylthiazol

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-4-phenylthiazole

024114-250MG
FLUO024114-250MGEA 48.8 EUR
FLUO024114-250MG
2-Chlor-4-phenylthiazol
2-Chloro-4-phenylthiazol
Formel: C₉H₆ClNS
Molecular Weight: 195.668 g/mol
MDL: MFCD00956371
CAS-Nummer: 1826-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR