2-Cyan-4,6-dichloranilin

Lieferant: FLUOROCHEM

024191-25G 024191-5G
FLUO024191-25GEA 180 EUR
FLUO024191-25G FLUO024191-5G
2-Cyan-4,6-dichloranilin
2-Cyan-4,6-dichloranilin
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187.03 g/mol
MDL: MFCD00158947
CAS-Nummer: 36764-94-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR