3-Brom-4-methoxybenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Brom-4-methoxybenzoesäurechlorid

024494-1G
FLUO024494-1GEA 405 EUR
FLUO024494-1G
3-Brom-4-methoxybenzoylchlorid
3-Brom-4-methoxybenzoylchlorid
Formel: C₈H₆BrClO₂
Molecular Weight: 249,49 g/mol
MDL: MFCD02628379
CAS-Nummer: 81324-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR