2-Chlor-5-hydroxypyridin

Lieferant: FLUOROCHEM
Achtung

024512-5G
FLUO024512-5GEA 60.9 EUR
FLUO024512-5G
2-Chlor-5-hydroxypyridin
2-Chlor-5-hydroxypyridin
Formel: C₅H₄ClNO
MDL: MFCD00234050
CAS-Nummer: 41288-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR