1-Brom-3-chlor-2-fluorbenzol

Lieferant: FLUOROCHEM

Synonyme: 3-Chloro-2-fluorobromobenzene

024515-5G 024515-25G
FLUO024515-5GEA 81.3 EUR
FLUO024515-5G FLUO024515-25G
1-Brom-3-chlor-2-fluorbenzol
1-Bromo-3-chloro-2-fluorobenzene
Formel: C₆H₃BrClF
MDL: MFCD03412182
CAS-Nummer: 144584-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR