3-Brom-2-chlortoluol

Lieferant: FLUOROCHEM

024558-25G
FLUO024558-25GEA 233 EUR
FLUO024558-25G
3-Brom-2-chlortoluol
3-Brom-2-chlortoluol
Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
MDL: MFCD07782165
CAS-Nummer: 97329-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR