1,3-Difluor-5-iodbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3,5-Difluoroiodobenzene

024566-25G
FLUO024566-25GEA 152 EUR
FLUO024566-25G
1,3-Difluor-5-iodbenzol
1,3-Difluoro-5-iodobenzene
Formel: C₆H₃F₂I
Molecular Weight: 239,99 g/mol
MDL: MFCD03094155
CAS-Nummer: 2265-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR