3-Fluor-2-methylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-2-methylbenzol-1-sulfonylchlorid, 3-Fluor-2-methylbenzolsulfochlorid, 3-Fluor-2-methylbenzolsulfonsäurechlorid

024625-1G
FLUO024625-1GEA 125 EUR
FLUO024625-1G
3-Fluor-2-methylbenzolsulfonylchlorid
3-Fluor-2-methylbenzolsulfonylchlorid
Formel: C₇H₆ClFO₂S
Molecular Weight: 208,64 g/mol
MDL: MFCD07782175
CAS-Nummer: 875166-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR