4-Fluor-3-methylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

024626-5G
FLUO024626-5GEA 461 EUR
FLUO024626-5G
4-Fluor-3-methylbenzolsulfonylchlorid
4-Fluor-3-methylbenzolsulfonylchlorid
Formel: C₇H₆ClFO₂S
Molecular Weight: 208,63 g/mol
MDL: MFCD01365818
CAS-Nummer: 629672-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR