5-Brom-2-chloranilin

Lieferant: FLUOROCHEM

024803-25G
FLUO024803-25GEA 96.3 EUR
FLUO024803-25G
5-Brom-2-chloranilin
5-Brom-2-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
MDL: MFCD06796283
CAS-Nummer: 60811-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR