(-)-Di-p-toluoyl-L-weinsäure

Lieferant: FLUOROCHEM

Synonyme: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

024870-100G
FLUO024870-100GEA 87.9 EUR
FLUO024870-100G
(-)-Di-p-toluoyl-L-weinsäure
(-)-Di-p-toluoyl-L-weinsäure
Formel: C₂₀H₁₈O₈
Molecular Weight: 386,36 g/mol
MDL: MFCD00064440
CAS-Nummer: 32634-66-5
EINECS: 251-131-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR