4-Chlor-2-fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Chlor-2-fluorbenzolboronsäure, (4-Chlor-2-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-2-fluorphenyl)boran

024877-5G
FLUO024877-5GEA 63.9 EUR
FLUO024877-5G
4-Chlor-2-fluorphenylboronsäure
4-Chlor-2-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
MDL: MFCD02684293
CAS-Nummer: 160591-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR