2-Chlor-4-(trifluormethoxy)phenol

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-2-hydroxy-5-(trifluormethoxy)benzol

024882-5G
FLUO024882-5GEA 264 EUR
FLUO024882-5G
2-Chlor-4-(trifluormethoxy)phenol
2-Chlor-4-(trifluormethoxy)phenol
Formel: C₇H₄ClF₃O₂
Molecular Weight: 212,56 g/mol
MDL: MFCD04972753
CAS-Nummer: 70783-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR