1-Brom-2,4-difluor-5-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 5-Brom-2,4-difluornitrobenzol

024967-25G
FLUO024967-25GEA 204 EUR
FLUO024967-25G
1-Brom-2,4-difluor-5-nitrobenzol
1-Bromo-2,4-difluoro-5-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237,99 g/mol
MDL: MFCD00129166
CAS-Nummer: 345-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR