5-Brom-2-nitrobenzoesäure

Lieferant: FLUOROCHEM

024975-5G
FLUO024975-5GEA 76.2 EUR
FLUO024975-5G
5-Brom-2-nitrobenzoesäure
5-Brom-2-nitrobenzoesäure
Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
MDL: MFCD00093013
CAS-Nummer: 6950-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR