3-Amino-1-methyl-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole

025069-25G
FLUO025069-25GEA 0 EUR
FLUO025069-25G
3-Amino-1-methyl-1H-pyrazol
3-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97,12 g/mol
MDL: MFCD00466340
CAS-Nummer: 1904-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR