3-Nitro-1H-pyrazol
Lieferant: FLUOROCHEM
026154-1G
FLUO026154-1GEA
31.1
EUR
FLUO026154-1G
3-Nitro-1H-pyrazol
3-Nitro-1H-pyrazole
Formel:
C₃H₃N₃O₂ Molecular Weight: 113,08 g/mol |
MDL:
MFCD00238787 CAS-Nummer: 26621-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...