4-Nitro-1H-pyrazol

Lieferant: FLUOROCHEM

026197-25G
FLUO026197-25GEA 102 EUR
FLUO026197-25G
4-Nitro-1H-pyrazol
4-Nitro-1H-pyrazole
Formel: C₃H₃N₃O₂
Molecular Weight: 113,0800 g/mol
MDL: MFCD00159626
CAS-Nummer: 2075-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR