1,3-Difluor-2-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 2,6-Difluoroiodobenzene

032511-5G
FLUO032511-5GEA 44.3 EUR
FLUO032511-5G
1,3-Difluor-2-iodbenzol
1,3-Difluoro-2-iodobenzene
Formel: C₆H₃F₂I
Molecular Weight: 239,99 g/mol
MDL: MFCD04039247
CAS-Nummer: 13697-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR