3-Fluor-2-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

032726-1G
FLUO032726-1GEA 90.9 EUR
FLUO032726-1G
3-Fluor-2-(trifluormethyl)pyridin
3-Fluoro-2-(trifluoromethyl)pyridine
Formel: C₆H₃F₄N
Molecular Weight: 165.09 g/mol
MDL: MFCD08437598
CAS-Nummer: 886510-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR