5-Brompicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

033053-25G
FLUO033053-25GEA 79.1 EUR
FLUO033053-25G
5-Brompicolinsäure
5-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
MDL: MFCD00234149
CAS-Nummer: 30766-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR