4-Fluor-3-methoxybenzolsulfonylchlorid

Lieferant: FLUOROCHEM

033071-5G
FLUO033071-5GEA 798 EUR
FLUO033071-5G
4-Fluor-3-methoxybenzolsulfonylchlorid
4-Fluor-3-methoxybenzolsulfonylchlorid
Formel: C₇H₆ClFO₃S
MDL: MFCD07368708
CAS-Nummer: 887266-97-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR