4-Fluor-3-(trifluormethyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

033095-5G
FLUO033095-5GEA 87.8 EUR
FLUO033095-5G
4-Fluor-3-(trifluormethyl)phenylboronsäure
4-Fluor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
MDL: MFCD07363787
CAS-Nummer: 182344-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR