2-Fluor-5-pyridinboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluorpyridin-5-boronsäure, 2-Fluor-5-pyridinylboronsäure

033100-1G
FLUO033100-1GEA 50 EUR
FLUO033100-1G
2-Fluor-5-pyridinboronsäure
2-Fluor-5-pyridinboronsäure
Formel: C₅H₅BFNO₂
Molecular Weight: 140,91 g/mol
MDL: MFCD03411559
CAS-Nummer: 351019-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR