2-Brom-6-fluorpyridin

Lieferant: FLUOROCHEM

033114-25G
FLUO033114-25GEA 165 EUR
FLUO033114-25G
2-Brom-6-fluorpyridin
2-Bromo-6-fluoropyridine
Formel: C₅H₃BrFN
MDL: MFCD03095258
CAS-Nummer: 144100-07-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR