5-Fluor-2-nitroanisol

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-2-methoxy-1-nitrobenzene

033120-25G
FLUO033120-25GEA 78.8 EUR
FLUO033120-25G
5-Fluor-2-nitroanisol
5-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077541
CAS-Nummer: 448-19-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR