3-Brompicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid

033124-5G
FLUO033124-5GEA 0 EUR
FLUO033124-5G
3-Brompicolinsäure
3-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
MDL: MFCD01320380
CAS-Nummer: 30683-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR