2-Chlor-4-methoxybenzoesäure
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-p-anissäure
033210-1G
FLUO033210-1GEA
51.8
EUR
FLUO033210-1G
2-Chlor-4-methoxybenzoesäure
2-Chloro-4-methoxybenzoic acid
Formel:
C₈H₇ClO₃ |
MDL:
MFCD00085943 CAS-Nummer: 21971-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...