3-Chlor-2-pyridincarbonitril

Lieferant: FLUOROCHEM

033256-5G
FLUO033256-5GEA 63.8 EUR
FLUO033256-5G
3-Chlor-2-pyridincarbonitril
3-Chlor-2-pyridincarbonitril
Formel: C₆H₃ClN₂
MDL: MFCD03788834
CAS-Nummer: 38180-46-0
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR