6-Aminopicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 6-Aminopyridine-2-carboxylic acid

033257-1G
FLUO033257-1GEA 31.1 EUR
FLUO033257-1G
6-Aminopicolinsäure
6-Aminopicolinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00233711
CAS-Nummer: 23628-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR