5-Brom-2-methylpyridin
Lieferant: FLUOROCHEM
Synonyme:
3-Brom-6-picolin, 5-Brom-2-picolin
033262-25G
FLUO033262-25GEA
104
EUR
FLUO033262-25G
5-Brom-2-methylpyridin
5-Brom-2-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol |
MDL:
MFCD00661170 CAS-Nummer: 3430-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...