3,3,3-Trifluorpropionitril

Lieferant: FLUOROCHEM

033300-5G
FLUO033300-5GEA 165 EUR
FLUO033300-5G
3,3,3-Trifluorpropionitril
3,3,3-Trifluorpropionitril
Formel: C₃H₂F₃N
MDL: MFCD07784277
CAS-Nummer: 20530-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR