3-Chlor-5-iodbenzoesäure

Lieferant: FLUOROCHEM

033365-5G
FLUO033365-5GEA 198 EUR
FLUO033365-5G
3-Chlor-5-iodbenzoesäure
3-Chlor-5-iodbenzoesäure
Formel: C₇H₄ClIO₂
Molecular Weight: 282,46 g/mol
MDL: MFCD00672966
CAS-Nummer: 289039-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR