3-Hydroxy-5-(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

033427-5G
FLUO033427-5GEA 375 EUR
FLUO033427-5G
3-Hydroxy-5-(trifluormethyl)benzoesäure
3-Hydroxy-5-(trifluoromethyl)benzoic acid
Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
MDL: MFCD07368783
CAS-Nummer: 328-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR