2-Amino-5-fluorbenzothiazol
Lieferant: FLUOROCHEM
Synonyme:
5-Fluorobenzo[d]thiazol-2-amine
033441-1G
FLUO033441-1GEA
110
EUR
FLUO033441-1G
2-Amino-5-fluorbenzothiazol
2-Amino-5-fluorobenzothiazole
Formel:
C₇H₅FN₂S Molecular Weight: 168,19 g/mol |
MDL:
MFCD05664560 CAS-Nummer: 20358-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...