2-Amino-3-brom-5-chlorpyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin

033564-5G
FLUO033564-5GEA 257 EUR
FLUO033564-5G
2-Amino-3-brom-5-chlorpyridin
2-Amino-3-brom-5-chlorpyridin
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
MDL: MFCD00955627
CAS-Nummer: 26163-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR