α,2-Dibrom-5-fluortoluol

Lieferant: FLUOROCHEM

Synonyme: 1-Bromo-2-(bromomethyl)-4-fluorobenzene, 2-Bromo-5-fluorobenzyl bromide

033733-25G
FLUO033733-25GEA 139 EUR
FLUO033733-25G
α,2-Dibrom-5-fluortoluol
α,2-Dibrom-5-fluortoluol
Formel: C₇H₅Br₂F
Molecular Weight: 267,92 g/mol
MDL: MFCD00042187
CAS-Nummer: 112399-50-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR