5-Chlor-2-fluorbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-4-fluor-3-(trifluormethyl)benzol, 4-Chlor-1-fluor-2-(trifluormethyl)benzol, 5-Chlor-2-fluorbenzotrifluorid

033737-5G
FLUO033737-5GEA 125 EUR
FLUO033737-5G
5-Chlor-2-fluorbenzotrifluorid
5-Chlor-2-fluorbenzotrifluorid
Formel: C₇H₃ClF₄
Molecular Weight: 198,55 g/mol
MDL: MFCD00060655
CAS-Nummer: 89634-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR