4-Fluor-2-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 5-Fluoro-2-hydroxybenzotrifluoride

033739-5G
FLUO033739-5GEA 240 EUR
FLUO033739-5G
4-Fluor-2-(trifluormethyl)phenol
4-Fluor-2-(trifluormethyl)phenol
Formel: C₇H₄F₄O
Molecular Weight: 180,1 g/mol
MDL: MFCD00061290
CAS-Nummer: 130047-19-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR