5-Fluor-2-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-2-hydroxybenzotrifluoride

033749-5G
FLUO033749-5GEA 257 EUR
FLUO033749-5G
5-Fluor-2-(trifluormethyl)phenol
5-Fluor-2-(trifluormethyl)phenol
Formel: C₇H₄F₄O
Molecular Weight: 180,1 g/mol
MDL: MFCD00236302
CAS-Nummer: 243459-91-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR