3,5-Difluor-4-methoxybenzoesäure

Lieferant: FLUOROCHEM

033760-1G
FLUO033760-1GEA 58.1 EUR
FLUO033760-1G
3,5-Difluor-4-methoxybenzoesäure
3,5-Difluoro-4-methoxybenzoic acid
Formel: C₈H₆F₂O₃
MDL: MFCD01090994
CAS-Nummer: 319-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR