4'-Hydroxy-3'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

033762-1G
FLUO033762-1GEA 237 EUR
FLUO033762-1G
4'-Hydroxy-3'-(trifluormethyl)acetophenon
4'-Hydroxy-3'-(trifluormethyl)acetophenon
Formel: C₉H₇F₃O₂
MDL: MFCD01091005
CAS-Nummer: 149105-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR