4-Brom-2-fluor-6-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

033800-5G
FLUO033800-5GEA 173 EUR
FLUO033800-5G
4-Brom-2-fluor-6-(trifluormethyl)anilin
4-Brom-2-fluor-6-(trifluormethyl)anilin
Formel: C₇H₄BrF₄N
CAS-Nummer: 875664-46-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR