2-Brom-6-nitrophenol

Lieferant: FLUOROCHEM

034301-25G
FLUO034301-25GEA 103 EUR
FLUO034301-25G
2-Brom-6-nitrophenol
2-Brom-6-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218,01 g/mol
MDL: MFCD02683216
CAS-Nummer: 13073-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR