Methyl-5-fluor-2-nitrobenzoat

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-nitrobenzoesäuremethylester

034754-25G
FLUO034754-25GEA 168 EUR
FLUO034754-25G
Methyl-5-fluor-2-nitrobenzoat
Methyl-5-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,14 g/mol
MDL: MFCD03425619
CAS-Nummer: 393-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR